Learn More
Nabertherm Steel Collecting Pan
For use with LE 2 furnace. Protects the furnace and provides easy loading.
Brand:  Nabertherm 691400626
Product Code. 15783309
  • 238.00€ / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Steel construction
  • For use with LE 2 model furnace
  • Dimensions (L x W x H): 110 x 170 x 20 mm
  • Maximum Temperature: 1100°C
Specifications
Specifications
Protects the furnace and provides easy loading