missing translation for 'onlineSavingsMsg'
Learn More

behr Labor-Technik™ Steam Distillation Unit With Automatic Addition

behr distillation units of the S series are the ideal supplement to the behr InKjel digestion systems. These are identical in the basic construction but differ in terms of operating comfort and degree of automation.

Brand:  behr Labor-Technik™ B00218025

 View more versions of this product

Product Code. 10345072

  • 16505.00€ / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket

Récupel-bijdrage uitgesloten
This item is not returnable. View return policy

Description

Description

  • Exemplary safety and reliability
    • Robust and insensitive casing made of polyurethane
      • Distillation time approx. 3 min. per sample
        • Detection limit 0.2 mg N
          • Recovery rate 99.5%
            • Reproducibility ± 1%
              • Adjustable steam generating capacity (10% - 100%)
                • Very easy, menu-guided operation with a single operation element (the behr one-button operation)
                  • Programmable reaction time
                    • Programmable distillation time
                      • USB interface
                        • Fill-level monitoring for canister set
                          • Practical quick clamping device
                            • Performance capacity and speed
                              • Practical and user friendly
                                • Exemplary safety and reliability
                                  • Robust and insensitive casing made of polyurethane
                                    • Extremely simple, menu-controlled operation of the programmable units via
                                      • one single operating element (behr one-button operation).
                                        • Menu navigation in country language
Specifications

Specifications

S1 Steam Distillation Unit
1700 W
Any Size Supply Container
1 Program
42 cm
9 A
34.5 cm
70 cm
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.