Learn More
IBA Lifesciences StarGate pPSG-IBA Acceptor Vectors for E. coli Expression
Product Code. 15653758 Shop All IBA Lifesciences Products
Click to view available options
Vector:
pPSG-IBA 105
pPSG-IBA 123
pPSG-IBA 164
pPSG-IBA 168
pPSG-IBA 23
pPSG-IBA 25
pPSG-IBA 3
pPSG-IBA 33
pPSG-IBA 5
pPSG-IBA 62
pPSG-IBA 63
pPSG-IBA 64
pPSG-IBA 65
pPSG-IBA wt1
pPSG-IBA103
pPSG-IBA143
pPSG-IBA145
pPSG-IBA162
pPSG-IBA167
pPSG-IBA35
pPSG-IBA43
pPSG-IBA45
Unit Size:
5µg
Product Code. 15653758
Brand: IBA Lifesciences 54205001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Achieves high-level expression in E. coli by the T7 promoter by providing specified genetic surroundings for optimal expression and purification of non-toxic proteins.
pPSG-IGA E. coli expression vectors:
pPSG-IBA vectors use the T7 promoter and T7 RNA polymerase for high-level transcription of the gene of interest.
  • As the T7 promoter is stronger than the tet promoter, pPSG-IBA vectors are recommended in cases where expression with the tet promoter does not lead to significant yields of the recombinant protein.
    • High-level expression in E. coli by bacteriophage T7 promoter
    • High-level transcription by T7 RNA polymerase in BL21 strains
    • High-level expression of non-toxic proteins
    • Induction by IPTG
    • Suitable for in vitro transcription/translation
    StarGate System
    IBA developed StarGate to provide a simple, fast and highly efficient cloning process for genes of interest (GOI) for use in functional expression, purification, detection, immobilization or separation of recombinant proteins:
    • Minimal extra modification of the gene due to short combinatorial sites
    • Inherent high-level cloning efficiency due to a directed reaction (no equilibrium)
    • GOIs are cloned to obtain a Donor Vector; 2 or more genes can be combined using Fusion Cloning
    • Genes are then transferred into Acceptor Vectors that provide optimal genetic surroundings
    • The final expression vector, the Destination Vector, is then placed into the respective host
  • Specifications
    Format Liquid
    For Use With (Equipment) StarGate™ System, E. Coli BL21(DE3)
    Inducing Agent E. coli host with T7 RNA polymerase gene (e.g. BL21(DE3))
    pH 8
    Promoter T7
    Product Type Acceptor Vector for E. coli expression
    Storage Buffer 20 μL TE buffer: 10 mm Tris-HCl, 1 mm EDTA / 5 μg
    Content And Storage 2°C to 8°C for frequent usage, -20°C for long-term storage
    Protein Tag Strep-tag for N-terminal
    Concentration 250 ng/μL
    Quantity 5 μg
    Vector pPSG-IBA 5
    Color Clear
    For Use With (Application) Cloning genes of interest for use in functional expression, purification, detection, immobilization or separation of recombinant proteins