Learn More
WTW™ Standard Calibration and Storage Sleeve for WTW™ Oxygen Meter
The calibration and storage containers for the WTW oxygen sensors are suitable for the standard conform calibration (DIN ISO 5814 or DIN ISO 17289: 2014-12) with water steam saturated air as well as for storage of the sensors to protect the sensitive membrane.
Brand:  WTW™ 205362
Product Code. 10519911
  • 224.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Specifications
Standard calibration and storage sleeve
Oxygen meter