Learn More
Hanna Instruments™ Thermistor Probe with Handle, Stainless Steel
Obtain highly accurate temperature readings of liquids, gases and/or semi-solid products. Hanna Instruments™ Thermistor Probe with Handle, Stainless Steel feature a handle designed for use with a thermistor thermometer.
Brand:  Hanna Instruments™ HI765A
 View more versions of this product
Product Code. 10534271
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
These thermistor probes feature a cable that extends 1m (3.3 ft.) from the handle to the meter connection. (Options with a 2m or 3m cable are also available.) With strong, corrosion-resistant, AISI 316 stainless steel construction, this probe is able to measure samples up to 150°C (302°F).
The HI765 series of PTC thermistor probes is pre-calibrated with a maximum error of ±0.2°C (±0.4°F), so recalibration is not necessary when probes are interchanged.
  • AISI 316 stainless steel probe with ABS handle
  • Applications: air and gas, air and liquid, and semi-solid options
  • NTC thermistor with 6 second response time, and pre-calibrated PTC thermistors with 8 second response time
Specifications
Specifications
Air and gas temperature
8.1 in.
3 m
100 mm
8 sec.
PTC
Yes
±0.2°C
9.94 ft.
3.9 in.
205 mm
Stainless Steel (AISI 316) Probe, ABS Handle
HI 765A Air Probe
Thermistor Probe
Thermistor thermometer