Learn More
Consort™ Stainless Steel Temperature Compensator
Brand:  Consort™ ST10N
Product Code. 11995498
  • 211.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Specifications
4 mm
Field use
Stainless Steel
Temperature Compensator
110 mm
-30°C to +130°C