Learn More
Aalborg™ Precision Valve for Stainless Steel Flowmeter
Aalborg™ Precision Valve for Stainless Steel Flowmeter is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
Brand:  Aalborg™ VM3-SV
 View more versions of this product
Product Code. 11778368
  • 214.00€ / Each
Explore more special offers
Description
Description
  • For high precision
  • Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
  • They may also be used as shut off valves.
  • Bodies of the valves are reinforced by structurally rigid metallic shells.
  • Shells are made of stainless steel, bushings are made of plated brass or 316 stainless steel.
Specifications
Specifications
1000 mL/min.
26mL/min.
150 mm Gas/Liquid Flowmeter
Standard Valve