Learn More
Aalborg™ Precision Valve for Stainless Steel Flowmeter
Aalborg™ Precision Valve for Stainless Steel Flowmeter is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
Brand: Aalborg™ VM2-SV
This item is not returnable.
View return policy
Description
- For high precision
- Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
- They may also be used as shut off valves.
- Bodies of the valves are reinforced by structurally rigid metallic shells.
- Shells are made of stainless steel, bushings are made of plated brass or 316 stainless steel.
Specifications
400 mL/min. | |
12mL/min. | |
150 mm Gas/Liquid Flowmeter | |
Standard Valve |