Learn More
Buerkle™ Stainless Steel Safety Jerrycans
Buerkle™ Stainless Steel Safety Jerrycans offer safety through flame protection and excess pressure valve.
708.00€ - 1120.00€
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | |||||
15395087
|
Buerkle™
2601-3005 |
5 L | Screw Cap, Pressure Relief Valve |
708.00€
Each |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||||
11951551
|
Buerkle™
2601-7010 |
10 L | Screw Cap |
713.40€
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
|||||
15358498
|
Buerkle™
2601-2010 |
10 L | Fine Dosage Spout, Pressure-relief Valve |
829.60€
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
15574429
|
Buerkle™
2601-3020 |
20 L | Screw Cap, Pressure Relief Valve |
1050.00€
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
16445079
|
Buerkle™
2601-2020 |
20 L | Fine Dosage Spout, Pressure-relief Valve, Ventilation |
1120.00€
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
|||||
11941551
|
Buerkle™
2601-3010 |
10 L | Screw Cap and Pressure Relief Valve |
729.33€
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
Description
- Handles made of solid stainless steel
- Seals made of PTFE
- High degree of chemical resistance
- Save space
Specifications
Stainless Steel |
Product Title
Buerkle™ Stainless Steel Safety Jerrycans