Learn More
Grant Instruments™ Stainless Steel Perforated Base Tray
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
104.94€ - 135.00€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11426417
|
Grant Instruments™
SBT14 |
DMU12 & SAP12 |
113.00€
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
11456417
|
Grant Instruments™
SBT28 |
DMU19, SAP18/26 |
120.00€
Each |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
11406417
|
Grant Instruments™
SBT6 |
DMU5, SAP2/5, JBN5 |
120.00€
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
11466417
|
Grant Instruments™
SBT36 |
SAP34 |
116.00€
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
15315832
|
Grant Instruments™
SBT12 |
LSB12 |
135.00€
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
15345832
|
Grant Instruments™
SBT26 |
OLS26/LSB18 |
134.00€
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
11416417
|
Grant Instruments™
SBT2 |
SAP2 |
104.94€
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
Description
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
- For use with SBB Aqua Plus and LSB Series Aqua Pro Series Unstirred Water Bath
- Choose from a vartiety of sizes.