missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVP Stainless Steel M-26XV Model Transilluminator
Can be used with any UVP imaging system
Brand: UVP 95-0413-02
This item is not returnable.
View return policy
Description
UVP's range of benchtop transilluminators can be used with any UVP imaging system. They use 8W tubes and have a specially constructed filter that provides the most uniform light emission possible from a tube based unit. The units have a stainless steel top that is easy to clean and a hinged UV blocking lid to protect the user. Single wavelength units have high, medium and low intensity settings that make the units suitable for both visualisation and band cutting. The dual and 3UV wavelength units have a single intensity setting. Single and dual wavelength benchtop transilluminator, dimensions 279mm x 356mm x 122mm3UV benchtop transilluminator, dimensions 279mm x 356mm x 137mmSpecifications
356 mm | |
279 mm | |
UVP Imaging System | |
230 V 50 Hz | |
M-26XV | |
50 Hz | |
302 nm |
122 mm | |
Single UV Transilluminator | |
250 x 260 mm | |
250 x 260 mm | |
230 V | |
High/Medium/Low | |
UV Benchtop |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction