Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Needle, Custom length, Point Style 2, 3, or 4
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
160.00€ - 316.00€
Specifications
Product Type | Luer Lock Custom Needle |
---|---|
Needle Point Style | 2, 3, or 4 |
For Use With (Equipment) | TLL Syringes and LT or TLL Connectors |
Connection Type | Standard Tapered |
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
11592435
|
Hamilton™
7749-01 |
10 G |
N/A
|
N/A | |||||
10388249
|
Hamilton™
7748-01 |
15 G |
N/A
|
N/A | |||||
11512445
|
Hamilton™
7749-03 |
12 G |
N/A
|
N/A | |||||
10546623
|
Hamilton™
7749-05 |
14 G |
N/A
|
N/A | |||||
10443305
|
Hamilton™
7749-04 |
13 G |
N/A
|
N/A | |||||
11592335
|
Hamilton™
7748-02 |
16 G |
N/A
|
N/A | |||||
10642203
|
Hamilton™
7748-18 |
22s G |
N/A
|
N/A | |||||
11542345
|
Hamilton™
7748-09 |
23 G |
N/A
|
N/A | |||||
11552345
|
Hamilton™
7748-10 |
24 G |
N/A
|
N/A | |||||
11582345
|
Hamilton™
7748-13 |
27 G |
N/A
|
N/A | |||||
11512355
|
Hamilton™
7748-19 |
26s G |
N/A
|
N/A | |||||
11572345
|
Hamilton™
7748-12 |
26 G |
N/A
|
N/A | |||||
10133882
|
Hamilton™
7748-14 |
28 G |
N/A
|
N/A | |||||
15139246
|
Hamilton™
7748-14 |
28 G |
N/A
|
N/A | |||||
10244502
|
Hamilton™
7747-01 |
33 G |
N/A
|
N/A | |||||
Description
- Available in various gauges
- Autoclavable
- Metal hubs manufactured from nickel plated brass
- Custom length
- Compatible with PTFE Luer Lock (TLL) syringes
- Design controlled by a series of universal standards which guarantees compatibility between manufacturers
- Provided in packs of 6
Specifications
Luer Lock Custom Needle | |
2, 3, or 4 | |
TLL Syringes and LT or TLL Connectors | |
Standard Tapered |