Learn More
Blanco™ Gastronorm Stainless Steel Lids with Handle
Product Code. 11963836
Click to view available options
Size:
GN 1/1
GN 1/2
GN 1/3
GN 1/4
GN 1/6
GN 2/3
Length (Metric):
176 mm
265 mm
325 mm
354 mm
530 mm
Width (Metric):
162 mm
176 mm
265 mm
325 mm
Unit Size:
Each
Product Code. 11963836
Brand: Blanco™ 550654
Only null left
Add to basket
Add to basket
Fits most Gastronorm containers, from 1/1 to 1/6. Blanco™ Gastronorm Stainless Steel Lids with Handle are dishwasher-safe, stackable lids made of AISI 304 stainless steel with ergonomically shaped handles to promote safe handling and easy access.
Gastronorm Stainless Steel Lids feature outstanding stability thanks to circumferential edge stamping. These rugged and dimensionally stable lids are easy to clean and offer optimum hygiene thanks to the smooth, non-porous surface.
  • First-class, corrosion-resistant finish with burr-free edges
  • Temperature-resistant from −40°C to +280°C
  • Tasteless and odorless stainless steel (AISI 304)
Specifications
Product Type Lid with Handle
Material Stainless Steel (304)
Certifications/Compliance European standard EN 631
For Use With (Equipment) Gastronome containers
Length (Metric) 265 mm
Quantity 1 Each
Shape Rectangle
Size GN 1/4
Temperature Range (Metric) −40°C to 280°C
Width (Metric) 162 mm