Learn More
Welch Ilmvac™ Stainless Steel KF Flange with Long Tubulation
Shop All Welch Ilmvac Products
missing translation for 'orderingAttributeHoverText'
Diameter (Metric) Inner:
16 mm
24 mm
40.5 mm
Diameter (Metric) Outer:
20 mm
28 mm
44.5 mm
missing translation for 'unitSize'
Each
Specifications
Specifications
| Diameter (Metric) Inner | 40.5 mm |
| Material | Stainless Steel |
| Diameter (Metric) Outer | 44.5 mm |
| Overall Length (Metric) | 5.8 cm |
| Packaging Quantity | 1 Each |
| Tubing Size | DN 40 |
| Product Type | KF Flange with Long Tubulation |