Learn More
Agencinox™ Stainless Steel Flat Bottom Dish
Use these dishes in decontamination services, operating rooms, surgeries, clinics and healthcare fields. Agencinox™ Stainless Steel Flat Bottom Dishes are ideal for holding dressings and other items which may be needed during surgical procedures.
Brand:  Agencinox™ 304
 View more versions of this product
Product Code. 11729503
  • 69.00€ / Pack of 5
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Flat bottom stainless steel lotion bowls
  • Autoclavable
  • Available in a variety of sizes
Specifications
Specifications
Flat
Stainless Steel
Dish
40 mm
20 mL
Round
20 mm