Learn More
Sampling Systems™ Stainless Steel Economy Scoops
Shop All Sampling Systems Products
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
120 mL
450 mL
1000 mL
1500 mL
2000 mL
missing translation for 'unitSize'
Each
Description
- Manufactured from spot welded 304 stainless steel
- With an attractive high polish
- Light duty construction
- Not suitable for GMP applications as there are crevices
Specifications
Specifications
| Capacity (Metric) | 2000 mL |
| Product Type | Economy Scoop |
| Finish | High polish |
| For Use With (Application) | General industrial use |
| Material | Stainless Steel (AISI 304) |