Learn More
Fisherbrand™ Stainless Steel Cylindrical Seamless Scoops
Product Code. 11588092 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
150 mL
500 mL
1000 mL
4000 mL
Unit Size:
Each
Product Code. 11588092
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Get optimum results weighing and handling samples and other laboratory materials with cylindrical scoops. Perfect for weighing anything from liquid to gems and other materials, Fisherbrand™ Stainless Steel Seamless Cylindrical Scoops are particularly suitable for pharmaceutical applications.
  • Stainless steel seamless cylindrical scoops 
  • Particularly suitable for pharmaceutical applications  
  • All surfaces are non-reacting 
  • Offer zero absorption suiting GMP requirements
  • Scratches within the scoop prevent cross contamination
Specifications
Autoclavable Autoclavable
Shape Cylindrical
Capacity (Metric) 1000 mL
Product Type Scoop
For Use With (Application) Suitable for Pharmaceutical Applications
Length (Metric) Handle 120 mm
Material Stainless Steel
Length (Metric) 340 mm