Learn More
Sampling Systems™ Stainless Steel Buckets with Skirt Band and Graduations
Sampling Systems™ Stainless Steel Buckets with Skirt Band and Graduations are made from heavy gauge, durable stainless steel and are ideal for hygienic applications.
108.00€ - 124.00€
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15859601
|
Sampling Systems™
A264-10 |
Capacity: 10L |
108.00€
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
15869601
|
Sampling Systems™
A264-12 |
Capacity: 12L |
117.00€
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||||
15879601
|
Sampling Systems™
A264-15 |
Capacity: 15L |
124.00€
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||||
Description
- Crevice free interior
- Graduated
- Supplied with skirt band
- Robust design
- 304 Stainless Steel