missing translation for 'onlineSavingsMsg'
Learn More

ssPA/Stringent Starvation Protein A Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-06079JF549

Product Code. 18517289

  • 407.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ssPA/Stringent Starvation Protein A Polyclonal antibody specifically detects ssPA/Stringent Starvation Protein A in E. coli samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ssPA/Stringent Starvation Protein A
Polyclonal
ELISA
ECK3218, Pog, ssp, stringent starvation protein A
Produced in rabbits immunized with purified, recombinant Escherichia coli ssPA/Stringent Starvation Protein A (Uniprot#: P0ACA3; Met 1-Ser 212).
0.1 mL
Primary
E. coli
Purified
ELISA
Janelia Fluor 549
50mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
944744
Store at 4C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.