Learn More
SREBF1 Rabbit, anti-Human, Clone: 1P4, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82684-3-RR-20UL
Description
The SREBF1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human SREBF1. This antibody recognizes human antigen. The SREBF1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| SREBF1 | |
| 1P4 | |
| bHLHd1, SREBF1, SREBP 1, SREBP 1c, SREBP1 | |
| 20 μL | |
| 6720 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 800 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |