missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR 11237, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Pan retinoid X receptor (RXR) agonist that is devoid of any RAR activity.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 380.48 |
Percent Purity | >98% |
Chemical Name or Material | SR 11237 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'