missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sphingosine Kinase Substrate
Click to view available options
Quantity:
400nmol
Unit Size:
Each
Description
Sphingosine substrate specifically optimized for use with the Sphingosine kinases (SPHK1 & SPHK2). For use with the Adapta™ Universal Kinase Assay (PV5099).
Specifications
Specifications
| Chromagen Quantity | 400nmol |
| For Use With (Application) | Pharma and biopharma, target and lead identification and validation, drug discovery and development, kinase biology, biochemical kinase assays, Adapta™ universal kinase assays |
| Quantity | 400nmol |
| Substrate Type | Kinase Substrate |
| Product Type | Sphingosine Kinase Substrate |
| Storage Requirements | –20°C |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction