missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Micro Float-A-Lyzer™ Ready-to-Use Small Volume Dialysis Device

Providing ultimate convenience, this preassembled device incorporates Spectrum's ultrapure Biotech Grade Cellulose Ester (CE) membrane tubing into an easy-to-use design making laboroatory microdialysis easier than ever

Brand:  Spectrum™ F235053

178.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12045625

  • 183.00€ / Pack of 12

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Providing ultimate convenience, this preassembled device incorporates Spectrum's ultrapure Biotech Grade Cellulose Ester (CE) membrane tubing into an easy-to-use design making laboroatory microdialysis easier than ever

Ready-to-Use and disposable, the Float-A-Lyzer G2 incorporates Biotech Grade Cellulose Ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use & provides ultimate convenience. Available in volume sizes: 1, 5, and 10 mL. The resealable screw cap is color-coded for 9 available MWCO's and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5 and 10mL sizes). The flotation ring (included) provides buoyancy to maximize dialysis efficiency.

  • Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis
  • Leak-proof Luer-Lok™ cap provides easy access with the included syringe for easy loading and sample retrieval
  • Units can also interlock for the simultaneous dialysis of multiple samples
  • Ready-to-use and complete
  • Easy syringe loading (no needle)
  • Total sample recovery
  • Ultrapure Biotech CE (no cleaning)
  • Largest range of MWCOs
  • Maximizes SA & minimizes hold-up volume

Specifications

Specifications

Micro Float-A-Lyzer Dialysis Device
Cellulose Ester
0.25 in.
1.7 in.
1.77 in.
100 to 200 μL
Polypropylene
1mL syringe
Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis
3.5 to 5 kDa
Polycarbonate
6.4 mm
44 mm
45 mm
Black
Luer lok
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.