Learn More
Spectrum™ Micro Float-A-Lyzer™ Ready-to-Use Small Volume Dialysis Device
Providing ultimate convenience, this preassembled device incorporates Spectrum's ultrapure Biotech Grade Cellulose Ester (CE) membrane tubing into an easy-to-use design making laboroatory microdialysis easier than ever
Brand:  Spectrum™ F235063
 View more versions of this product
Product Code. 12035625
  • 183.00€ / Pack of 12
Explore more special offers
Description
Description
Providing ultimate convenience, this preassembled device incorporates Spectrum's ultrapure Biotech Grade Cellulose Ester (CE) membrane tubing into an easy-to-use design making laboroatory microdialysis easier than ever
Ready-to-Use and disposable, the Float-A-Lyzer G2 incorporates Biotech Grade Cellulose Ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use & provides ultimate convenience. Available in volume sizes: 1, 5, and 10 mL. The resealable screw cap is color-coded for 9 available MWCO's and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5 and 10mL sizes). The flotation ring (included) provides buoyancy to maximize dialysis efficiency.
  • Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis
  • Leak-proof Luer-Lok™ cap provides easy access with the included syringe for easy loading and sample retrieval
  • Units can also interlock for the simultaneous dialysis of multiple samples
  • Ready-to-use and complete
  • Easy syringe loading (no needle)
  • Total sample recovery
  • Ultrapure Biotech CE (no cleaning)
  • Largest range of MWCOs
  • Maximizes SA & minimizes hold-up volume
Specifications
Specifications
Micro Float-A-Lyzer Dialysis Device
Cellulose Ester
0.25 in.
2.4 in.
1.77 in.
400 to 500 μL
Polypropylene
1mL syringe
Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis
0.5 to 1 kDa
Polycarbonate
6.4 mm
63 mm
45 mm
Orange
Luer lok