Learn More
Biochrom Spectophotometer Calibration Set
missing translation for 'orderingAttributeHoverText'
Type:
Spectophotometer Calibration Set
missing translation for 'unitSize'
Each
Description
NIST Calibrated with easy to use software
Specifications
Specifications
| Description | Spectophotometer Calibration Set |
| Type | Spectophotometer Calibration Set |
| For Use With (Equipment) | Biochrom™ Ultrospec 2100/7000/8000/9000, Libra S11/12/21/22/50/60/70/80, and BioDrop Spectrophotometers |