Learn More
Eppendorf™ Spare Rotor Lids
Designed for use with Eppendorf Rotors
Brand: Eppendorf™ 5424703003
260.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Spare Rotor Lid
- Available in a variety of sizes
- Aerosol tight
- For use with liquid handling applications
Specifications
Lid | |
Rotor FA-45-24-11 |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%