Learn More
Notrax Sof-Tred™ Microcellular Vinyl Foam with Pebble Pattern, Gray
Notrax Sof-Tred™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit.
Brand: Notrax 411C0348GY
This item is not returnable.
View return policy
Description
- Pebble pattern provides non-directional traction and is easy to clean
- Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
- All four sides are bevelled and sealed to minimize tripping hazards
Specifications
Gray | |
1830 cm | |
Industrial and Commercial Matting |
60 ft. | |
Microcellular Vinyl | |
122 cm |