missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tert-pentyloxide, 95%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
Beschreibung
Sodium tert-pentyloxide is a effective base for the alkylation of ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium tert-pentyloxide is a effective base for the alkylation of ketones.
Solubility
Reacts with water.
Notes
Air and Moisture Sensitive, store away form air and water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Sodium tert-pentyloxide is a effective base for the alkylation of ketones.
Solubility
Reacts with water.
Notes
Air and Moisture Sensitive, store away form air and water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spezifikation
Odor | Odorless |
Quantity | 25 g |
UN Number | UN3206 |
Beilstein | 3983987 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 110.13 |
Percent Purity | 95% |
Chemical Name or Material | Sodium tert-pentyloxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'