missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution, 0.1M, Chem-Lab
Click to view available options
Quantity:
1 L
5 L
Description
Sodium hydroxide is used in many industries, mostly as a strong chemical base in the manufacture of pulp and paper, textiles, drinking water, soaps and detergents and as a drain cleaner.
Specifications
Specifications
pH | >13 |
Color | Colorless |
Boiling Point | 100°C |
Concentration | 0.1M |
Density | 1.01g/mL |
Solubility Information | Water Soluble |
Quantity | 1 L |
Formula Weight | 40.00g/mol |
Concentration or Composition (by Analyte or Components) | 4g NaOH per 1L H2O |
Normality | 0.1 N (±0.0002, 20°C) |
Show More |
Safety and Handling
Product Identifier
- Sodium hydroxide 0.1 mol/l
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Sodium hydroxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction