Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Culture Tube
Product Code. 11803370
Click to view available options
Length (Metric):
40 mm
50 mm
55 mm
70 mm
120 mm
125 mm
150 mm
Diameter (Metric):
8 mm
9.5 mm
10.5 mm
11 mm
12 mm
16 mm
Unit Size:
Pack of 200
Pack of 250
Pack of 500
Product Code. 11803370
Brand: DWK Life Sciences A412012000711
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use the Soda Lime Glass Culture Tube to view cell cultures in the laboratory. This disposable soda lime glass rim top tube is plain (unlabeled) and features a sturdy, uniform bottom.
  • Rimmed top
  • Unlableled
  • Clear soda lime glass
  • Uniform bottom
  • Disposable
TRUSTED_SUSTAINABILITY
Specifications
Design Type Round bottom
Length (Metric) 120 mm
Packaging 500Pack
Diameter (Metric) 12 mm
Product Type Test Tube