Learn More
Brannan™ General Purpose Specific Gravity Hydrometers Series 50
Product Code. 12811448
Click to view available options
Specific Gravity Range:
0.7 to 0.75 g/mL
0.750 to 0.800 g/mL
0.800 to 0.850 g/mL
0.850 to 0.900 g/mL
0.9 to 0.95 g/mL
0.950 to 1.000 g/mL
1.000 to 1.050 g/mL
1.050 to 1.100 g/mL
1.10 to 1.15 g/mL
1.150 to 1.200 g/mL
1.20 to 1.25 g/mL
1.250 to 1.300 g/mL
1.300 to 1.350 g/mL
1.35 to 1.40 g/mL
1.40 to 1.45 g/mL
1.45 to 1.50 g/mL
1.500 to 1.550 g/mL
1.550 to 1.600 g/mL
Unit Size:
Each
Product Code. 12811448
Brand: Brannan™ 10/901/0
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Shot loaded specific gravity hydrometers for a variety of surface tension ranges.
  • Shot loaded
  • Length 190 mm
  • Scale subdivisions 0.002
  • Accuracy ±0.002 specific gravity
Specifications
Quantity 1 Ea.
Type Specific Gravity Hydrometer
Increments 0.002 g/mL
Certifications/Compliance UKAS
Length (Metric) 190 mm
Specific Gravity Range 0.7 to 0.75 g/mL