missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SMO/CEN7q (Texas Red/FITC) FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
1515.00€ - 2495.00€
Specifications
Content And Storage | Store at 4°C in the dark. |
---|---|
Description | Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique. |
Format | Tube |
Label or Dye | FITC, Texas Red |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023245
|
Abnova™
FG0153.200uL |
200 μL |
2495.00€
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16013245
|
Abnova™
FG0153.100uL |
100 μL |
1515.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
Store at 4°C in the dark. | |
Tube | |
Fluorescent In Situ Hybridization (Cell) | |
7q32.3/7q11.21 | |
SMO/CEN7q FISH Probe | |
Human |
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique. | |
FITC, Texas Red | |
DAPI Counterstain (1500 ng/mL ) 250 uL | |
Approx. 290kb/800kb | |
Genomic DNA | |
Liquid |