Learn More
Eppendorf™ SmartBlock DWP1000
Click to view available options
For Use With (Equipment):
Deepwell plates, 96/1,000 μL
Description
- For Eppendorf Deepwell Plate 96/1000 μL
- Optimized block design supports optimal temperature transfer within your sample to receive excellent results
- Eppendorf SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
- Every SmartBlock is individually inspected in production for temperature accuracy and is delivered includes serial-number-specific certificate for documentation purpose
- Insulated thermoblocks for ergonomic operation - you won't burn your fingers (PhysioCare Concept)
- Automatic block recognition for safe handling
- Eppendorf QuickRelease™ technology enables fast and simple block exchange - no need for tools
- Limited to ThermoMixer C and ThermoStat C
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
For Use With (Equipment) | Deepwell plates, 96/1,000 μL |
Type | Block |
Includes | Lid |