Learn More
Brady™ Small Core Pre-printed GHS CLP Labels
Supplied with pre-printed red diamonds, ready for you to print the required warning, compliant to the latest regulations.
Brand:  Brady™ 622324
 View more versions of this product
Product Code. 30041480
  • 118.00€ / Pack of 300
Explore more special offers
Description
Description
For you to print the required warning, compliant to the latest regulations.
Specifications
Specifications
UL Recognized
Permanent Acrylic
Red on White
Polypropylene
Thermal Transfer
Non-sterile
Hazard communication identification
1
R6000HF
300 Stickers
75.00 mm (W) x 110.00 mm (H)
-20°C to +100°C
0.079 mm
4.33 in.
2.95 in.
75.00 mm
Pre-printed Desktop Printer Labels for CLP-GHS Hazardous Substances
None
Gloss
Front
Labels
4 Red Diamonds in Square Formation per Sticker
BBP11, BBP12, PR Plus, THT 25 mm Core Printers
-20°C
300 Labels/Roll
Rectangle
-4°F to 212°F
0.0031 in.
75.00 mm (W) x 110.00 mm (H)
110.00 mm
75.00 mm
75.00 mm