Learn More
Brady™ Small Core Pre-printed GHS CLP Labels
Supplied with pre-printed red diamonds, ready for you to print the required warning, compliant to the latest regulations.
Brand:  Brady™ 622339
 View more versions of this product
Product Code. 30041476
  • 118.00€ / Pack of 200
Explore more special offers
Description
Description
For you to print the required warning, compliant to the latest regulations.
Specifications
Specifications
Pre-printed Desktop Printer Labels for CLP-GHS Hazardous Substances
None
Matte
Front
Labels
3 Stacked Red Diamonds in Pyramid Formation per Sticker
BBP11, BBP12, PR Plus, THT 25 mm Core Printers
-40°C
200 Labels/Roll
Rectangle
-40°F to +122°F
0.0032 in.
105.00 mm (W) x 155.00 mm (H)
155.00 mm
105.00 mm
105.00 mm
Permanent Acrylic
Red on White
Paper
Thermal Transfer
Non-sterile
Hazard communication identification
1
R7950
200 Stickers
105.00 mm (W) x 155.00 mm (H)
-40°C to +50°C
0.081 mm
6.1 in.
4.13 in.
105.00 mm