missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Small Core Pre-printed GHS CLP Labels

Supplied with pre-printed red diamonds, ready for you to print the required warning, compliant to the latest regulations.

Brand:  Brady™ 622315

 View more versions of this product

Product Code. 30041452

  • 110.00€ / Pack of 400

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

For you to print the required warning, compliant to the latest regulations.

Specifications

Specifications

Pre-printed Desktop Printer Labels for CLP-GHS Hazardous Substances
None
Matte
Front
Labels
55.00 mm
BBP11, BBP12, PR Plus, THT 25 mm Core Printers
-40°C
400 Labels/Roll
Rectangle
-40°F to +122°F
0.0032 in.
55.00 mm (W) x 85.00 mm (H)
85.00 mm
55.00 mm
3 Stacked Red Diamonds in Pyramid Formation per Sticker
Permanent Acrylic
Red on White
Paper
Thermal Transfer
Non-sterile
Hazard communication identification
1
R7950
400 Stickers
55.00 mm (W) x 85.00 mm (H)
-40°C to +50°C
0.081 mm
3.35 in.
2.17 in.
55.00 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions