missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Small Core Pre-printed GHS CLP Labels
Supplied with pre-printed red diamonds, ready for you to print the required warning, compliant to the latest regulations.
Brand: Brady™ 622336
This item is not returnable.
View return policy
Description
For you to print the required warning, compliant to the latest regulations.
Specifications
UL Recognized | |
Permanent Acrylic | |
Red on White | |
Polypropylene | |
Thermal Transfer | |
Non-sterile | |
Hazard communication identification | |
1 | |
R6000HF | |
200 Stickers | |
105.00 mm (W) x 155.00 mm (H) | |
-20°C to +100°C | |
0.079 mm | |
6.1 in. | |
4.13 in. | |
105.00 mm |
Pre-printed Desktop Printer Labels for CLP-GHS Hazardous Substances | |
None | |
Gloss | |
Front | |
Labels | |
4 Red Diamonds in Square Formation per Sticker | |
BBP11, BBP12, PR Plus, THT 25 mm Core Printers | |
-20°C | |
200 Labels/Roll | |
Rectangle | |
-4°F to 212°F | |
0.0031 in. | |
105.00 mm (W) x 155.00 mm (H) | |
155.00 mm | |
105.00 mm | |
105.00 mm |