Learn More
Invitrogen™ SlowFade™ Gold Antifade Mountant
Description
SlowFade Gold Antifade Mountant is a liquid mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides. It contains chemical components designed to protect fluorescent dyes from fading (photobleaching) during fluorescence microscopy experiments. SlowFade Gold Antifade Mountant is glycerol-based, with no mixing or curing required, which makes it ideal for immediate viewing of the sample. It comes ready-to-use—just apply a drop to the sample, add a coverslip, and image. It is available with or without DAPI nuclear stain.
Key attributes:
• Protects dyes from fading during imaging
• Ready-to-use liquid, ideal for immediate sample viewing
• Mounted samples are stable for weeks
• Maintains signal strength—little to no quenching
• Ideal for Alexa Fluor dyes
SlowFade Gold Antifade Mountant is not recommended for mounting samples containing fluorescent proteins, such as GFP. For superior antifade protection of fluorescent proteins and fluorescent dyes in cultured cells or thin tissue slices, SlowFade Diamond Antifade Mountant is recommended. For high resolution or to image thicker tissue or 3D cell culture with a focal depth of 0–500 μm, try SlowFade Glass Antifade Mountant.
- Environmental benefits include:
- Recycled Content
- Non-Toxic
- Source Reduction
Specifications
Specifications
Content And Storage | Storage at room temperature is recommended but can also be stored frozen (-5° to -30°C). Protect from light. |
For Use With (Equipment) | Microscope |
Product Line | SlowFade™ |
Product Type | Mounting Media |
Quantity | 10 mL |
Solution Type | Anti-fade Mountant |
Green Features | Less hazardous, sustainable packaging |
Shipping Condition | Room Temperature |
Form | Liquid |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24158Expires: 2025-10-31
Days
42Hours
2Minutes
18Seconds
55 -
Multiple Manufacturers
Save Up to 63%
Big savings on advanced technologies and materials testing.Promo Code: 24141Expires: 2025-12-14
New Offer Save Up to 63%