missing translation for 'onlineSavingsMsg'
Learn More

3M™ Safety-Walk™ Slip Resistant Conformable Tape, Series 500

Product Code. 17118670
Click to view available options
Color:
Black
Yellow
Dimensions (L x W):
18 x 0.051 m
18.3 x 0.102 m
18.3 x 0.305 m
Unit Size:
1 roll
Pack of 2
This item is not returnable. View return policy

Product Code. 17118670

Brand: 3M™ Safety-Walk™ 7000126117

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Ideal for use on diamond grade safety plates, steps, and other irregular surfaces. Mineral coated and slip resistant, with aluminum foil backing. Conforms to irregular surfaces and around edges.

  • Anti-slip tape for irregular or contoured surfaces in light to heavy shoe-traffic areas
  • Aluminum backing conforms to corners, curves, and irregular surfaces like diamond plating, riveted plates, and ladder rungs
  • Suitable for application in dry, wet or oily environments
  • Aggressive, high performance adhesive forms an excellent bond for a secure hold
  • Tapes and treads resist wrinkling, curling, tearing, shrinking, and lifting
  • Safety yellow color for high visibility and easy identification of hazard areas
  • Custom shapes and sizes are also available
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W) 18.3 x 0.102 m
For Use With (Application) Light to Heavy Traffic Areas
Length (Metric) 18.3 m
Packaging Quantity 1 Ea.
Width (English) 4 in.
Width (Metric) 0.102 m
Adhesive Type Pressure Sensitive Adhesive
Color Black
Type Floor Tape
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.