missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC7A7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542432
This item is not returnable.
View return policy
Description
Peptide sequence: WGTLVQDIFT YAKVLALIAV IVAGIVRLGQ GASTHFENSF EGSSFAVGDI Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 79%; Pig: 93%; Rabbit: 100%; Rat: 79%
SLC7A7 Polyclonal antibody specifically detects SLC7A7 in Human samples. It is validated for Western Blot.Specifications
SLC7A7 | |
Polyclonal | |
Unconjugated | |
SLC7A7 | |
AI790233; amino acid transporter SLC7A7 y+LAT1; LAT3; LPI; monocyte amino acid permease 2; MOP-2; my+lat1; Slc7a7; solute carrier family 7 (amino acid transporter light chain, y+L system), member 7; solute carrier family 7 (cationic amino acid transporter, y+ system), member 7; solute carrier family 7 member 7; y(+)L-type amino acid transporter 1; Y+L amino acid transporter 1; Y+L amino acid transporter 1; solute carrier family 7 member 7; y+LAT1; y+LAT-1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9056 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UM01 | |
SLC7A7 | |
Synthetic peptide directed towards the middle region of human SLC7A7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |