missing translation for 'onlineSavingsMsg'
Learn More

SLC7A7 Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Brand:  Invitrogen PA5143049

Product Code. 17898142

  • 425.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

The protein encoded by this gene is the light subunit of a cationic amino acid transporter. This sodium-independent transporter is formed when the light subunit encoded by this gene dimerizes with the heavy subunit transporter protein SLC3A2. This transporter is found in epithelial cell membranes where it transfers cationic and large neutral amino acids from the cell to the extracellular space. Defects in this gene are a cause of lysinuric protein intolerance (LPI). Several transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC7A7
Polyclonal
Unconjugated
SLC7A7
AI790233; amino acid transporter SLC7A7 y+LAT1; LAT3; LPI; monocyte amino acid permease 2; MOP-2; my+lat1; Slc7a7; solute carrier family 7 (amino acid transporter light chain, y+L system), member 7; solute carrier family 7 (cationic amino acid transporter, y+ system), member 7; solute carrier family 7 member 7; y(+)L-type amino acid transporter 1; Y+L amino acid transporter 1; Y+L amino acid transporter 1; solute carrier family 7 member 7; y+LAT1; y+LAT-1
Goat
Ammonium sulfate precipitation
RUO
9056
-20° C, Avoid Freeze/Thaw Cycles
IgG
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q9UM01
SLC7A7
Peptide with sequence SQPEVETSPLGD-C.
100 μg
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions