missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC7A7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA557574
This item is not returnable.
View return policy
Description
Immunogen sequence: DSTEYEVASQ PEVETSPLGD GASPGPEQVK LKK Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 58%.
The protein encoded by this gene is the light subunit of a cationic amino acid transporter. This sodium-independent transporter is formed when the light subunit encoded by this gene dimerizes with the heavy subunit transporter protein SLC3A2. This transporter is found in epithelial cell membranes where it transfers cationic and large neutral amino acids from the cell to the extracellular space. Defects in this gene are a cause of lysinuric protein intolerance (LPI). Several transcript variants encoding the same protein have been found for this gene.Specifications
SLC7A7 | |
Polyclonal | |
Unconjugated | |
SLC7A7 | |
AI790233; amino acid transporter SLC7A7 y+LAT1; LAT3; LPI; monocyte amino acid permease 2; MOP-2; my+lat1; Slc7a7; solute carrier family 7 (amino acid transporter light chain, y+L system), member 7; solute carrier family 7 (cationic amino acid transporter, y+ system), member 7; solute carrier family 7 member 7; y(+)L-type amino acid transporter 1; Y+L amino acid transporter 1; Y+L amino acid transporter 1; solute carrier family 7 member 7; y+LAT1; y+LAT-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9056 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.15 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UM01 | |
SLC7A7 | |
Recombinant protein corresponding to Human SLC7A7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only