Learn More
Witeg™ Simple Distillation Kit
Product Code. 11857730
Click to view available options
Capacity (Metric):
250mL
Unit Size:
Each
Product Code. 11857730
Brand: Witeg™ A1100201
Simple distillation kit, complete with all components necessary for smaller scale distillation.
  • Ideal for easy, smaller scale distllation processes
  • Includes boiling flask, column, condenser, spigot, clamps
Specifications
Includes Boiling flask, column, condenser tube, spigot, clamps
Type Distillation Kit
Capacity (Metric) 250mL