missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Silylating MSTFA

Product Code. 10471142 Shop All Macherey-Nagel Products
Click to view available options
Quantity:
1 mL
Unit Size:
20mL
This item is not returnable. View return policy

Product Code. 10471142

Brand: Macherey-Nagel™ 701270.201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Item Description Silylating MSTFA
Quantity 1 mL
compliance-icons compliance-icons
Product Identifier
  • MSTFA, 20x1 mL
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Supplemental information
  • MIXTURE LIST-Contains : N -methyl-N-trimethylsilyl-trifluoroacetamide (MSTFA)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.