Learn More
Fisherbrand™ General Purpose Silicone Sheet
An excellent choice for applications requiring good sealing properties at high and low temperatures. Fisherbrand™ General Purpose Silicone Sheets are also suitable for applications requiring direct food contact.
Brand: Fisherbrand™ 3753752
This item is not returnable.
View return policy
Description
- Available plain or adhesive backed
- Suitable for use at -40° to +200°C and +230°C short-term
- Can be supplied in sheet form or converted into strips, seals, washers, gaskets and fabricated articles
Specifications
2 x 360 x 360mm |