Learn More
JG Finneran Associates™ Silicone Septa
Brand: JG Finneran Associates™ 604060-10
This item is not returnable.
View return policy
Specifications
PTFE | |
Silicone | |
For Screw Thread Closures | |
10-425 |
Red | |
Yes | |
1000/Pk. |