Learn More
Borer Chemie™ Silicone-free Grease Tube
Standard quality for desiccators, ground joints, stopcocks and apparatuses. Borer Chemie™ Silicone-Free Grease Tubes are suitable for sealing and lubricating fixed and rotary ground glass joints and for greasing fittings made of plastics and metal.
Brand:  Borer Chemie™ DGLR.0060
 View more versions of this product
Product Code. 11915756
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Excellent thermal stability
  • Available in standard quality (vacuum tight up to 300°C) and high-vacuum quality (up to 400°C)
  • Constant lubricating consistency between -40°C and +250°C
  • No dripping point
  • Chemically stable against most acids, alkalis and gases
  • Insoluble in water and low molecular mono- and polyhydric alcohols
Specifications
Specifications
Standard
60 g