missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica for MPLC, MP
Shop All MP Biomedicals Products
Click to view available options
:
MP Silica for MPLC; 500 g
Description
Produced from the identical raw material used for MP EcoChrom™ Silica for open column chromatography. Therefore, they show the same sorption behavior and allow easy switching between the methods.
Specifications
Specifications
| Boiling Point | 2230°C |
| Melting Point | 1710°C |
| CAS | 7631-86-9 |
| pH | 6.5 to 7.0 |
| Quantity | 500 g |
| Molecular Formula | O2Si |
| MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
| InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
| SMILES | O=[Si]=O |
| IUPAC Name | silanedione |
| Show More |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction