missing translation for 'onlineSavingsMsg'
Learn More

Silica for HPLC, MP

Product Code. 17807044
Click to view available options
MP Silica for HPLC, 10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Silica for HPLC's very closely controlled particle range guarantees dense column packings, high plate numbers and relatively low back pressure

Weight: 0.4 to 0.5 g/mL
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 2230°C
Melting Point 1710°C
CAS 7631-86-9
Color White
pH 7
Physical Form Powder
Quantity 10 g
Molecular Formula O2Si
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name silanedione
Molecular Weight (g/mol) 60.08
Loss on Drying 3.50%
Density 0.3 g/mL
Chemical Name or Material Silicon dioxide
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'