Learn More
BRAND™ SILBERBRAND Automatic Burette with PTFE Stockcock
Product Code. 10471141 Shop All Brand Products
Click to view available options
Includes:
2000 mL bottle
Without bottle
Unit Size:
Each
Product Code. 10471141
Brand: BRAND™ 22332
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
SILBERBRAND Automatic Burette equipped with an i-stopcock and lateral stopcock.
  • Error limits according to class B, DIN EN ISO 385
  • Interchangeable keys
  • Calibrated to deliver (TD, Ex)
  • With glass-key in the titration and intermediate stopcock
  • Automatic zeroing
  • Intermediate stopcock to recirculate residual liquid
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 25 mL
Graduations 0.1 mL
Includes 2000 mL bottle
Stopcock i-stopcock, amber lateral stopcock, PTFE
Warranty 12 months